Assay Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199228.2 | NP_001186157.1 | 4 | 649 | 98 | |
| GenBank mRNA | HM991500.1 | - | 4 | 653 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNHIT3| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199228.2 | NP_001186157.1 | 4 | 649 | 98 | |
| GenBank mRNA | HM991500.1 | - | 4 | 653 | 98 | |
Set Membership: |
|