Assay Details
| Gene Symbol: RFT-II | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082402.1 | NP_001075871.1 | 1744 | 93 | ||
| GenBank mRNA | X80225.1 | - | 1744 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFT-II| Gene Symbol: RFT-II | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082402.1 | NP_001075871.1 | 1744 | 93 | ||
| GenBank mRNA | X80225.1 | - | 1744 | 93 | ||
Set Membership: |
|