Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342829.1 | XP_017198318.1 | 5 - 6 | 1153 | 68 | |
| XM_017342830.1 | XP_017198319.1 | 3 - 4 | 896 | 68 | ||
| XM_017342831.1 | XP_017198320.1 | 5 - 6 | 936 | 68 | ||
| XM_017342832.1 | XP_017198321.1 | 4 - 5 | 1070 | 68 | ||
| XM_017342833.1 | XP_017198322.1 | 4 - 5 | 1070 | 68 | ||
| XM_017342834.1 | XP_017198323.1 | 4 - 5 | 1070 | 68 | ||
More Information
Set Membership: |
|

MBD5
Genomic Map