Assay Details
| Gene Symbol: MAGED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272658.2 | XP_008270880.1 | 9 - 10 | 1339 | 60 | |
| XM_008272659.1 | XP_008270881.1 | 9 - 10 | 1335 | 60 | ||
| XM_008272660.2 | XP_008270882.1 | 9 - 10 | 1339 | 60 | ||
More Information
Set Membership: |
|

MAGED2
Genomic Map