Assay Details
| Gene Symbol: LOC100341681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257045.2 | XP_008255267.1 | 6 - 7 | 769 | 62 | |
| XM_017342096.1 | XP_017197585.1 | 6 - 7 | 769 | 62 | ||
| XM_017342097.1 | XP_017197586.1 | 6 - 7 | 769 | 62 | ||
| XM_017342098.1 | XP_017197587.1 | 6 - 7 | 769 | 62 | ||
| XM_017342099.1 | XP_017197588.1 | 6 - 7 | 769 | 62 | ||
| XM_017342100.1 | XP_017197589.1 | 6 - 7 | 769 | 62 | ||
| XM_017342101.1 | XP_017197590.1 | 6 - 7 | 769 | 62 | ||
More Information
Set Membership: |
|

LOC100341681
Genomic Map