Assay Details
| Gene Symbol: GEM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710692.3 | XP_002710738.2 | 4 - 5 | 815 | 67 | |
| XM_008255749.2 | XP_008253971.1 | 4 - 5 | 772 | 67 | ||
More Information
Set Membership: |
|

GEM
Genomic Map