Assay Details
| Gene Symbol: CDK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712792.3 | XP_002712838.1 | 4 - 5 | 965 | 72 | |
| XM_008259887.2 | XP_008258109.1 | 4 - 5 | 965 | 72 | ||
| XM_017343459.1 | XP_017198948.1 | 4 - 5 | 965 | 72 | ||
More Information
Set Membership: |
|

CDK8
Genomic Map