Assay Details
| Gene Symbol: EIF4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275238.2 | XP_008273460.1 | 643 | 63 | ||
| XM_017337525.1 | XP_017193014.1 | 643 | 63 | |||
| XM_017337526.1 | XP_017193015.1 | 643 | 63 | |||
More Information
Set Membership: |
|

EIF4E2
Genomic Map