Assay Details
| Gene Symbol: SELENOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256960.1 | NP_001243889.1 | 4 - 5 | 372 | 90 | |
| GenBank mRNA | DN883503.1 | - | 4 - 5 | 372 | 90 | |
| EB373960.1 | - | 4 - 5 | 347 | 90 | ||
| EL341919.1 | - | 4 - 5 | 358 | 90 | ||
More Information
Set Membership: |
|

SELENOF
Genomic Map