Assay Details
| Gene Symbol: RPS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285834.1 | NP_001272763.1 | 1 | 193 | 75 | |
| GenBank mRNA | DN883835.1 | - | 1 | 95 | 75 | |
| EL341892.1 | - | 1 | 194 | 75 | ||
More Information
Set Membership: |
|

RPS7
Genomic Map