Assay Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297476.1 | NP_001284405.1 | 3 - 4 | 283 | 80 | |
| XM_008259649.2 | XP_008257871.1 | 4 - 5 | 713 | 80 | ||
| XM_017343249.1 | XP_017198738.1 | 4 - 5 | 713 | 80 | ||
| XM_017343250.1 | XP_017198739.1 | 4 - 5 | 713 | 80 | ||
| GenBank mRNA | HQ738490.1 | - | 3 - 4 | 283 | 80 | |
More Information
Set Membership: |
|

CLEC1B
Genomic Map