Assay Details
| Gene Symbol: AKR1C1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297497.1 | NP_001284426.1 | 90 | 86 | ||
| XM_008248595.2 | XP_008246817.1 | 86 | ||||
| GenBank mRNA | AB854082.1 | - | 90 | 86 | ||
More Information
Set Membership: |
|

AKR1C1L
Genomic Map