Assay Details
| Gene Symbol: IL-15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301260.1 | NP_001288189.1 | 4 - 5 | 808 | 102 | |
| GenBank mRNA | JX271583.1 | - | 4 - 5 | 808 | 102 | |
More Information
Set Membership: |
|

IL-15L
Genomic Map