Assay Details
| Gene Symbol: CCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322432.1 | NP_001309361.1 | 2 - 3 | 487 | 67 | |
| GenBank mRNA | KU987815.1 | - | 2 - 3 | 181 | 67 | |
More Information
Set Membership: |
|

CCL11
Genomic Map