Assay Details
| Gene Symbol: IL18RAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329068.1 | NP_001315997.1 | 8 - 9 | 1130 | 66 | |
| XM_008253197.2 | XP_008251419.2 | 9 - 10 | 1436 | 66 | ||
| GenBank mRNA | JF271681.1 | - | 8 - 9 | 1208 | 66 | |
| JF271682.1 | - | 8 - 9 | 1130 | 66 | ||
| JF271683.1 | - | 7 - 8 | 886 | 66 | ||
More Information
Set Membership: |
|

IL18RAP
Genomic Map