Assay Details
| Gene Symbol: INHBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329069.1 | NP_001315998.1 | 1 - 2 | 392 | 68 | |
| GenBank mRNA | KC831577.1 | - | 1 - 2 | 392 | 68 | |
More Information
Set Membership: |
|

INHBA
Genomic Map