Assay Details
| Gene Symbol: LOC100349423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329074.1 | NP_001316003.1 | 1 - 2 | 93 | 102 | |
| XM_002716840.3 | XP_002716886.2 | 2 - 3 | 207 | 102 | ||
| XM_008266951.2 | XP_008265173.1 | 2 - 3 | 209 | 102 | ||
More Information
Set Membership: |
|

LOC100349423
Genomic Map