Assay Details
| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001349481.1 | NP_001336410.1 | 15 - 16 | 1733 | 96 | |
| XM_017348448.1 | XP_017203937.1 | 6 - 7 | 800 | 96 | ||
More Information
Set Membership: |
|

CC2D2B
Genomic Map