Assay Details
| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001349481.1 | NP_001336410.1 | 4 - 5 | 379 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CC2D2B| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001349481.1 | NP_001336410.1 | 4 - 5 | 379 | 95 | |
Set Membership: |
|