Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710636.3 | XP_002710682.1 | 5 - 6 | 926 | 66 | |
| XM_017341399.1 | XP_017196888.1 | 4 - 5 | 827 | 66 | ||
More Information
Set Membership: |
|

RDH10
Genomic Map