Assay Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710062.3 | XP_002710108.1 | 2 - 3 | 313 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SF3B6| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710062.3 | XP_002710108.1 | 2 - 3 | 313 | 102 | |
Set Membership: |
|