Assay Details
| Gene Symbol: TBX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713752.3 | XP_002713798.1 | 2 - 3 | 841 | 59 | |
| XM_017344423.1 | XP_017199912.1 | 2 - 3 | 841 | 59 | ||
More Information
Set Membership: |
|

TBX20
Genomic Map