Assay Details
| Gene Symbol: ME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708650.3 | XP_002708696.1 | 2 - 3 | 481 | 57 | |
| XM_008263027.2 | XP_008261249.1 | 3 - 4 | 568 | 57 | ||
| XM_017344170.1 | XP_017199659.1 | 2 - 3 | 485 | 57 | ||
| XM_017344172.1 | XP_017199661.1 | 2 - 3 | 481 | 57 | ||
More Information
Set Membership: |
|

ME3
Genomic Map