Assay Details
| Gene Symbol: LOC100350425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267462.2 | XP_008265684.1 | 9 - 10 | 2621 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100350425| Gene Symbol: LOC100350425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267462.2 | XP_008265684.1 | 9 - 10 | 2621 | 95 | |
Set Membership: |
|