Assay Details
| Gene Symbol: LOC100350425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267462.2 | XP_008265684.1 | 3 - 4 | 1564 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100350425| Gene Symbol: LOC100350425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267462.2 | XP_008265684.1 | 3 - 4 | 1564 | 109 | |
Set Membership: |
|