Assay Details
| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712700.2 | XP_002712746.1 | 5 - 6 | 1374 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYT10| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712700.2 | XP_002712746.1 | 5 - 6 | 1374 | 64 | |
Set Membership: |
|