Assay Details
| Gene Symbol: LOC100338355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248511.2 | XP_008246733.1 | 182 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008248511.2 transcripts.
Set Membership: |
|

LOC100338355
Genomic Map