Assay Details
| Gene Symbol: CCDC110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250689.2 | XP_008248911.2 | 5 - 6 | 437 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC110| Gene Symbol: CCDC110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250689.2 | XP_008248911.2 | 5 - 6 | 437 | 95 | |
Set Membership: |
|