Assay Details
| Gene Symbol: PIGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715414.3 | XP_002715460.1 | 1 - 2 | 104 | 60 | |
| XM_008264061.2 | XP_008262283.1 | 1 - 2 | 104 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002715414.3, XM_008264061.2 transcripts.
Set Membership: |
|

PIGC
Genomic Map