Assay Details
| Gene Symbol: COMMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723470.3 | XP_002723516.1 | 102 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002723470.3 transcripts.
Set Membership: |
|

COMMD5
Genomic Map