Assay Details
| Gene Symbol: TMEFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712353.3 | XP_002712399.1 | 2 - 3 | 923 | 71 | |
| XM_008258851.2 | XP_008257073.1 | 2 - 3 | 924 | 71 | ||
More Information
Set Membership: |
|

TMEFF2
Genomic Map