Assay Details
| Gene Symbol: PCDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712982.3 | XP_002713028.1 | 1 - 2 | 589 | 85 | |
| XM_002712983.3 | XP_002713029.1 | 1 - 2 | 588 | 85 | ||
| XM_008260042.2 | XP_008258264.1 | 1 - 2 | 589 | 85 | ||
| XM_008260043.2 | XP_008258265.1 | 1 - 2 | 588 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712982.3, XM_002712983.3, XM_008260042.2, XM_008260043.2 transcripts.
Set Membership: |
|

PCDH9
Genomic Map