Assay Details
| Gene Symbol: KCTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261166.2 | XP_008259388.2 | 2 - 3 | 1601 | 58 | |
| XM_017344093.1 | XP_017199582.1 | 2 - 3 | 629 | 58 | ||
| XM_017344094.1 | XP_017199583.1 | 2 - 3 | 1301 | 58 | ||
| XM_017344095.1 | XP_017199584.1 | 2 - 3 | 198 | 58 | ||
More Information
Set Membership: |
|

KCTD1
Genomic Map