Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718517.3 | XP_002718563.1 | 7 - 8 | 1208 | 79 | |
| XM_008270219.2 | XP_008268441.1 | 6 - 7 | 1136 | 79 | ||
| XM_017348429.1 | XP_017203918.1 | 5 - 6 | 1064 | 79 | ||
More Information
Set Membership: |
|

LGI1
Genomic Map