Assay Details
| Gene Symbol: LYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710497.3 | XP_002710543.1 | 8 - 9 | 999 | 62 | |
| XM_002710498.3 | XP_002710544.1 | 8 - 9 | 936 | 62 | ||
More Information
Set Membership: |
|

LYN
Genomic Map