Assay Details
| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262652.2 | XP_008260874.1 | 4 - 5 | 501 | 91 | |
| XM_008262653.2 | XP_008260875.1 | 4 - 5 | 507 | 91 | ||
More Information
Set Membership: |
|

POLR1H
Genomic Map