Assay Details
| Gene Symbol: TRIM25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271258.2 | XP_008269480.1 | 2 - 3 | 1057 | 76 | |
| XM_017349123.1 | XP_017204612.1 | 2 - 3 | 1064 | 76 | ||
More Information
Set Membership: |
|

TRIM25
Genomic Map