Assay Details
| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250698.2 | XP_008248920.1 | 1 - 2 | 191 | 58 | |
| XM_008250702.2 | XP_008248924.1 | 1 - 2 | 187 | 58 | ||
| XM_008250705.2 | XP_008248927.1 | 1 - 2 | 189 | 58 | ||
More Information
Set Membership: |
|

PDLIM3
Genomic Map