Assay Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262268.2 | XP_008260490.1 | 1 - 2 | 206 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008262268.2 transcripts.
Set Membership: |
|

SREK1IP1
Genomic Map