Assay Details
| Gene Symbol: ADORA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250163.2 | XP_008248385.1 | 721 | 104 | ||
| XM_008250164.2 | XP_008248386.1 | 3612 | 104 | |||
| XM_008250165.2 | XP_008248387.1 | 3742 | 104 | |||
More Information
Set Membership: |
|

ADORA2A
Genomic Map