Assay Details
| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249471.2 | XP_008247693.1 | 845 | 93 | ||
| XM_008249472.2 | XP_008247694.1 | 908 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLIC2| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249471.2 | XP_008247693.1 | 845 | 93 | ||
| XM_008249472.2 | XP_008247694.1 | 908 | 93 | |||
Set Membership: |
|