Assay Details
| Gene Symbol: PPIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258714.2 | XP_008256936.1 | 4 - 5 | 361 | 70 | |
| XM_008258716.2 | XP_008256938.1 | 5 - 6 | 530 | 70 | ||
| XM_008258717.2 | XP_008256939.1 | 4 - 5 | 466 | 70 | ||
| XM_008258718.2 | XP_008256940.1 | 4 - 5 | 489 | 70 | ||
| XM_017342915.1 | XP_017198404.1 | 5 - 6 | 434 | 70 | ||
| XM_017342916.1 | XP_017198405.1 | 5 - 6 | 497 | 70 | ||
| XM_017342917.1 | XP_017198406.1 | 4 - 5 | 244 | 70 | ||
More Information
Set Membership: |
|

PPIG
Genomic Map