Assay Details
| Gene Symbol: SLC25A38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713101.3 | XP_002713147.1 | 1 - 2 | 321 | 55 | |
| XM_008260273.2 | XP_008258495.1 | 2 - 3 | 236 | 55 | ||
More Information
Set Membership: |
|

SLC25A38
Genomic Map