Assay Details
| Gene Symbol: AHSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709692.3 | XP_002709738.1 | 2 - 3 | 385 | 62 | |
| XM_008254360.2 | XP_008252582.1 | 2 - 3 | 749 | 62 | ||
More Information
Set Membership: |
|

AHSA2
Genomic Map