Assay Details
| Gene Symbol: CADM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267104.2 | XP_008265326.2 | 9 - 10 | 1699 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CADM2| Gene Symbol: CADM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267104.2 | XP_008265326.2 | 9 - 10 | 1699 | 56 | |
Set Membership: |
|