Assay Details
| Gene Symbol: ARL4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271553.2 | XP_008269775.1 | 1 - 2 | 133 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008271553.2 transcripts.
Set Membership: |
|

ARL4D
Genomic Map