Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719738.3 | XP_002719784.1 | 3 - 4 | 763 | 81 | |
| XM_017349540.1 | XP_017205029.1 | 3 - 4 | 765 | 81 | ||
| XM_017349541.1 | XP_017205030.1 | 3 - 4 | 767 | 81 | ||
| XM_017349542.1 | XP_017205031.1 | 3 - 4 | 863 | 81 | ||
| XM_017349543.1 | XP_017205032.1 | 2 - 3 | 547 | 81 | ||
More Information
Set Membership: |
|

RNF34
Genomic Map