Assay Details
| Gene Symbol: PCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249169.2 | XP_008247391.1 | 220 | 99 | ||
| XM_017337981.1 | XP_017193470.1 | 1075 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCP2| Gene Symbol: PCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249169.2 | XP_008247391.1 | 220 | 99 | ||
| XM_017337981.1 | XP_017193470.1 | 1075 | 99 | |||
Set Membership: |
|