Assay Details
| Gene Symbol: C14H3orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716273.3 | XP_002716319.2 | 2 - 3 | 257 | 113 | |
| XM_017346865.1 | XP_017202354.1 | 2 - 3 | 434 | 113 | ||
More Information
Set Membership: |
|

C14H3orf33
Genomic Map