Assay Details
| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344681.1 | XP_017200170.1 | 14 | 3228 | 105 | |
| XM_017344682.1 | XP_017200171.1 | 13 | 3155 | 105 | ||
| XM_017344683.1 | XP_017200172.1 | 12 | 2620 | 105 | ||
| XM_017344684.1 | XP_017200173.1 | 12 | 2995 | 105 | ||
| XM_017344685.1 | XP_017200174.1 | 11 | 2503 | 105 | ||
More Information
Set Membership: |
|

RANBP3L
Genomic Map